Skip to Content
Merck
All Photos(2)

Key Documents

850185P

Avanti

08:0 PI(4,5)P2

Avanti Research - A Croda Brand

Synonym(s):

PIP2[4′,5′](8:0/8:0); 110665

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$185.00
500 μG
$736.00

$185.00


Estimated to ship on11 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$185.00
500 μG
$736.00

About This Item

Empirical Formula (Hill Notation):
C25H58N3O19P3
CAS Number:
Molecular Weight:
797.66
UNSPSC Code:
51191904
NACRES:
NA.25

$185.00


Estimated to ship on11 April 2025


Request a Bulk Order

description

1,2-dioctanoyl-sn-glycero-3-phospho-(1′-myo-inositol-4′,5′-bisphosphate) (ammonium salt)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850185P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850185P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](OP(O)([O-])=O)[C@H](OP([O-])(O)=O)[C@@H](O)[C@H]1O)=O)(OC(CCCCCCC)=O)COC(CCCCCCC)=O.[NH4+].[NH4+].[NH4+]

InChI

1S/C25H49O19P3.3H3N/c1-3-5-7-9-11-13-18(26)39-15-17(41-19(27)14-12-10-8-6-4-2)16-40-47(37,38)44-23-20(28)21(29)24(42-45(31,32)33)25(22(23)30)43-46(34,35)36;;;/h17,20-25,28-30H,3-16H2,1-2H3,(H,37,38)(H2,31,32,33)(H2,34,35,36);3*1H3/t17-,20-,21+,22+,23-,24-

InChI key

FRWPUPUJRUQWKA-OZTQGENFSA-N

General description

Although PI(4,5)P2 is a minor component of cell membranes, it plays a critical role as a substrate for a number of important signaling proteins. PI(4,5)P2 is an intermediate in the IP3/DAG pathway where it is hydrolyzed by phospholipase C to liberate the second messengers, inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG). PI(4,5)P2 is also a substrate for PI 3-kinase where it is phosphorylated to PI(3,4,5)P3, an activator of downstream signaling components such as the protein kinase AKT. The dioctanoyl derivative of PI(4,5)P2 is more water soluble than its long chain counterparts.

Application

08:0 PI(4,5)P2 is suitable for use to identify its binding site in transient receptor potential vanilloid 4 (TRPV4 N) terminus.[1] It has been used in in vitro binding assay to determine the binding of G-protein coupled receptor kinases (GRKs) and β2-adrenergic receptor (β2AR).[2]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850185P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850185P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural Basis of TRPV4 N Terminus Interaction with Syndapin/PACSIN1-3 and PIP2
Goretzki B, et al.
Structure, 26(12), 1583-1593 (2018)
Peter Y Mercredi et al.
Journal of molecular biology, 428(8), 1637-1655 (2016-03-20)
Assembly of HIV-1 particles is initiated by the trafficking of viral Gag polyproteins from the cytoplasm to the plasma membrane, where they co-localize and bud to form immature particles. Membrane targeting is mediated by the N-terminally myristoylated matrix (MA) domain
Structural and functional analysis of a beta2-adrenergic receptor complex with GRK5
Komolov KE, et al.
Cell, 169(3), 407-421 (2017)
Tao Liang et al.
The Journal of biological chemistry, 289(9), 6028-6040 (2014-01-17)
In β-cells, syntaxin (Syn)-1A interacts with SUR1 to inhibit ATP-sensitive potassium channels (KATP channels). PIP2 binds the Kir6.2 subunit to open KATP channels. PIP2 also modifies Syn-1A clustering in plasma membrane (PM) that may alter Syn-1A actions on PM proteins
Scott B Hansen et al.
Nature, 477(7365), 495-498 (2011-08-30)
The regulation of ion channel activity by specific lipid molecules is widely recognized as an integral component of electrical signalling in cells. In particular, phosphatidylinositol 4,5-bisphosphate (PIP(2)), a minor yet dynamic phospholipid component of cell membranes, is known to regulate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service