Skip to Content
Merck
All Photos(1)

Key Documents

890705P

Avanti

16:0-18:1 EPC (Cl Salt)

Avanti Research - A Croda Brand 890705P, powder

Synonym(s):

1-palmitoyl-2-oleoyl-sn-glycero-3-ethylphosphocholine (chloride salt)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$278.00
25 MG
$352.00

$278.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$278.00
25 MG
$352.00

About This Item

Empirical Formula (Hill Notation):
C44H87NO8PCl
CAS Number:
Molecular Weight:
824.59
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$278.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (890705P-10mg)
pkg of 1 × 25 mg (890705P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890705P

application(s)

advanced drug delivery

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)OCC.[Cl-]

InChI

1S/C44H87NO8P.ClH/c1-7-10-12-14-16-18-20-22-23-25-27-29-31-33-35-37-44(47)53-42(41-52-54(48,50-9-3)51-39-38-45(4,5)6)40-49-43(46)36-34-32-30-28-26-24-21-19-17-15-13-11-8-2;/h22-23,42H,7-21,24-41H2,1-6H3;1H/q+1;/p-1/b23-22-;/t42-,54?;/m1./s1

InChI key

SXRDCCSKGWBZFF-ISBKSCLYSA-M

General description

1-palmitoyl-2-oleoyl-sn-glycero-3-ethylphosphocholine (16:0-18:1 EPC) is cationic lipid containing an ethyl group attached to the phosphate moiety through an ester bond and has a choline headgroup.[1]

Application

1-palmitoyl-2-oleoyl-sn-glycero-3-ethylphosphocholine (chloride salt) (16:0-18:1 EPC (Cl Salt)) might be used:
  • as a cationic lipid to explore the change in organization and dynamics of a membrane-bound fluorescent probe in host membranes of varying charge[1]
  • in the preparation of small unilamellar cationic liposomes [2]
  • in a novel albumin-associated lipoplex formulation to evaluate the antitumoral efficacy of immuno-gene therapy and “suicide” gene therapy[3]

Biochem/physiol Actions

1-palmitoyl-2-oleoyl-sn-glycero-3-ethylphosphocholine (16:0-18:1 EPC) is a potential transfection agent, which is also similar to the natural lipids.[1]

Packaging

5 mL Clear Glass Sealed Ampule (890705P-10mg)
5 mL Clear Glass Sealed Ampule (890705P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Passadouro et al.
International journal of nanomedicine, 9, 3203-3217 (2014-07-26)
Pancreatic ductal adenocarcinoma (PDAC) is a highly aggressive and mortal cancer, characterized by a set of known mutations, invasive features, and aberrant microRNA expression that have been associated with hallmark malignant properties of PDAC. The lack of effective PDAC treatment
H Faneca et al.
Biochimica et biophysica acta, 1768(5), 1093-1102 (2007-02-14)
In the present work, we used a novel albumin-associated lipoplex formulation, containing the cationic lipid 1-palmitoyl-2-oleoyl-sn-glycero-3-ethylphosphocholine (EPOPC) and cholesterol (Chol), to evaluate the antitumoral efficacy of two gene therapy strategies: immuno-gene therapy, mediated by IL-12 gene expression, and "suicide" gene
Modulation of fluorophore environment in host membranes of varying charge
Kelkar DA, et al.
Journal of Fluorescence, 13(6), 459-466 (2003)
Wei Zong et al.
Langmuir : the ACS journal of surfaces and colloids, 34(23), 6874-6886 (2018-05-20)
The success of nanoparticulate formulations in drug delivery depends on various aspects including their toxicity, internalization, and intracellular location. Vesicular assemblies consisting of phospholipids and amphiphilic block copolymers are an emerging platform, which combines the benefits from liposomes and polymersomes
Li Wang et al.
Molecular pharmaceutics, 4(4), 615-623 (2007-04-06)
To date, the primary approach to improving the transfection properties of cationic lipids has been the synthesis of new kinds of cationic amphipaths. Recently, however, it was found that combining two cationic lipid derivatives having the same head group but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service