Skip to Content
Merck
All Photos(1)

Key Documents

890717O

Avanti

14:1 EPC (Tf Salt)

Avanti Research - A Croda Brand 890717O

Synonym(s):

1,2-dimyristoleoyl-sn-glycero-3-ethylphosphocholine (Tf salt)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$278.00

$278.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$278.00

About This Item

Empirical Formula (Hill Notation):
C39H73F3NO11PS
CAS Number:
Molecular Weight:
852.03
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$278.00


Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 10 mg (890717O-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890717O

lipid type

transfection
cationic lipids

shipped in

dry ice

storage temp.

−20°C

General description

Ethylphosphocholine (EPC) are amphiphilic cationic lipids containing hydrophilic headgroup and lipophilic tail.[1]

Biochem/physiol Actions

1,2-dimyristoleoyl-sn-glycero-3-ethylphosphocholine (14:1 EPC) is less toxic and is biodegradable. It is known to possess efficient DNA transfection activity.[1] 14:1 EPC is very closely identical to the natural triacylglyceride lipids.[2]

Packaging

5 mL Clear Glass Sealed Ampule (890717O-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differentially charged hollow core/shell lipid-polymer-lipid hybrid nanoparticles for small interfering RNA delivery.
Jinjun Shi et al.
Angewandte Chemie (International ed. in English), 50(31), 7027-7031 (2011-06-24)
Mikhail A Galkin et al.
Journal of visualized experiments : JoVE, (134) (2018-04-24)
Detergents are indispensable for delivery of membrane proteins into 30-100 nm small unilamellar vesicles, while more complex, larger model lipid bilayers are less compatible with detergents. Here we describe a strategy for bypassing this fundamental limitation using fusogenic oppositely charged
Li Wang et al.
Molecular pharmaceutics, 4(4), 615-623 (2007-04-06)
To date, the primary approach to improving the transfection properties of cationic lipids has been the synthesis of new kinds of cationic amphipaths. Recently, however, it was found that combining two cationic lipid derivatives having the same head group but
Rumiana Koynova et al.
The journal of physical chemistry. B, 111(27), 7786-7795 (2007-06-19)
Some mixtures of two cationic lipids including phospholipid compounds (O-ethylphosphatidylcholines) as well as common, commercially available cationic lipids, such as dimethylammonium bromides and trimethylammonium propanes, deliver therapeutic DNA considerably more efficiently than do the separate molecules. In an effort to
Rumiana Koynova et al.
Molecular pharmaceutics, 6(3), 951-958 (2009-04-04)
Synthetic cationic lipids are widely used components of nonviral gene carriers, and the factors regulating their transfection efficiency are the subject of considerable interest. In view of the important role that electrostatic interactions with the polyanionic nucleic acids play in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service