Skip to Content
Merck
All Photos(1)

Key Documents

890702P

Avanti

16:0 EPC (Cl Salt)

Avanti Research - A Croda Brand 890702P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-ethylphosphocholine (chloride salt)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$278.00

$278.00


Estimated to ship on31 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$278.00

About This Item

Empirical Formula (Hill Notation):
C42H85NO8PCl
CAS Number:
Molecular Weight:
798.55
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$278.00


Estimated to ship on31 May 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (890702P-10mg)
pkg of 1 × 100 mg (890702P-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand 890702P

lipid type

cationic lipids
transfection

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)OCC.[Cl-]

InChI

1S/C42H85NO8P.ClH/c1-7-10-12-14-16-18-20-22-24-26-28-30-32-34-41(44)47-38-40(39-50-52(46,48-9-3)49-37-36-43(4,5)6)51-42(45)35-33-31-29-27-25-23-21-19-17-15-13-11-8-2;/h40H,7-39H2,1-6H3;1H/q+1;/p-1/t40-,52?;/m1./s1

InChI key

XXTKXMLARLHZSM-WGVGMMHHSA-M

General description

16:0 EPC (1,2-dipalmitoyl-sn-glycero-3-ethylphosphocholine) is a saturated phospholipid[1] with choline as its cationic group.[2] It exists in the gel phase at physiological temperature.[3]

Application

16:0 EPC (Cl Salt) has been used in liposome preparation.[2] It has also been used as a component of cationic lipid mixture.[4]

Biochem/physiol Actions

16:0 EPC has the ability to transfect DNA into cells.[3]

Packaging

5 mL Clear Glass Sealed Ampule (890702P-100mg)
5 mL Clear Glass Sealed Ampule (890702P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of lipoplex structure and lipid phase changes
Liposomes, 399-423 (2010)
Biocompatible cationic lipids for the formulation of liposomal DNA vectors
Montis C, et al.
Soft Matter, 10(24), 4287-4297 (2014)
Effects of cis-and trans-unsaturated lipids on an interdigitated membrane
Smith EA, et al.
Biophysical Chemistry, 190, 1-7 (2014)
Mixtures of cationic lipid O-ethylphosphatidylcholine with membrane lipids and DNA: Phase diagrams
Koynova R and MacDonald RC
Biophysical Journal, 85(4), 2449-2465 (2003)
Chu Wang et al.
Langmuir : the ACS journal of surfaces and colloids, 36(6), 1615-1622 (2020-01-23)
Body fluids flow all over the body and affect the biological processes at biointerfaces. To simulate such a case, sum frequency generation (SFG) vibrational spectroscopy and a self-designed microfluidic chip were combined together to investigate the interaction between a pH-responsive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service