Skip to Content
Merck
All Photos(1)

Key Documents

870700P

Avanti

Coenzyme A (free acid)

Avanti Research - A Croda Brand 870700P, powder

Synonym(s):

Coenzyme A (free acid)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$191.00
25 MG
$261.00

$191.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$191.00
25 MG
$261.00

About This Item

Empirical Formula (Hill Notation):
C21H36N7O16P3S
CAS Number:
Molecular Weight:
767.53
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$191.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (870700P-10mg)
pkg of 1 × 25 mg (870700P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870700P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCS)=O)=O)(C(C)(COP(O)(OP(O)(OC[C@H]([C@H]1OP(O)(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H]

InChI

1S/C21H36N7O16P3S/c1-21(2,16(31)19(32)24-4-3-12(29)23-5-6-48)8-41-47(38,39)44-46(36,37)40-7-11-15(43-45(33,34)35)14(30)20(42-11)28-10-27-13-17(22)25-9-26-18(13)28/h9-11,14-16,20,30-31,48H,3-8H2,1-2H3,(H,23,29)(H,24,32)(H,36,37)(H,38,39)(H2,22,25,26)(H2,33,34,35)

InChI key

RGJOEKWQDUBAIZ-UHFFFAOYSA-N

General description

Coenzyme A is structurally composed of 2-aminoethanethiol, pantothenic acid and phosphorylated adenosine phosphate.[1]

Biochem/physiol Actions

Coenzyme A is a cofactor involved in acyl group transfer. It is essential for carbohydrate, lipid and protein metabolism. Coenzyme A acts as a cofactor for vitamin B coenzymes.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (870700P-10mg)
5 mL Amber Glass Screw Cap Vial (870700P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

General, Organic, and Biological Chemistry, 783-783 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service