Skip to Content
Merck
All Photos(1)

Key Documents

860720P

Avanti

Lyso iGB3 (synthetic)

powder

Synonym(s):

Isoglobotriaosylsphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$1,040.00

$1,040.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 μG
$1,040.00

About This Item

Empirical Formula (Hill Notation):
C36H67NO17
CAS Number:
Molecular Weight:
785.91
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$1,040.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

package of 1 × 500 μg (860720P-500UG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

General description

Isoglobotriaosylsphingosine (Lyso iGB3) is a truncated form of iGb3. It is a β-linked glycolipid.[1] Isogloboside 3 (iGb3) is a glycosphingolipid.[2]

Biochem/physiol Actions

Isogloboside 3 (iGb3) acts as an endogenous ligand for natural killer T (NKT) cells.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860720P-500UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and biological evaluation of alpha-galactosylceramide (KRN7000) and isoglobotrihexosylceramide (iGb3)
Xia C, et al.
Bioorganic & Medicinal Chemistry Letters, 16(8), 2195-2199 (2006)
Humans lack iGb3 due to the absence of functional iGb3-synthase: implications for NKT cell development and transplantation
Christiansen D, et al.
PLoS Biology, 6(7) (2008)
Psychosine variants as antigens for natural killer T cells
Deng S, et al.
Chemical Science, 8(3), 2204-2208 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service