Skip to Content
Merck
All Photos(1)

Documents

860722P

Avanti

C17:0 iGB3 (synthetic)

powder

Synonym(s):

C17:0 Isoglobotriaosylceramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H99NO18
CAS Number:
Molecular Weight:
1038.35
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

99% (TLC)

form

powder

packaging

package of 1 × 500 μg (860722P-500UG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

General description

Isoglobotriaosylceramide (iGB3) is a glycosphingolipid. It is produced in the lysosomal compartment by antigen presenting cells in mammals.

Biochem/physiol Actions

Isoglobotriaosylceramide (iGB3) acts as an antigen and is involved in activating natural killer T (NKT) cells. It is also implicated as a carbohydrate ligand for several receptors.

Packaging

5 mL Clear Glass Sealed Ampule (860722P-500UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Humans lack iGb3 due to the absence of functional iGb3-synthase: implications for NKT cell development and transplantation
Christiansen D, et al.
PLoS Biology, 6(7) (2008)
Sensitive quantitation of isoglobotriaosylceramide in the presence of isobaric components using electrospray ionization-ion trap mass spectrometry
Li Y, et al.
Glycobiology, 18(2), 166-176 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service