Skip to Content
Merck
All Photos(1)

Key Documents

860670P

Avanti

Sphingosine (d14:1)

Avanti Research - A Croda Brand 860670P, powder

Synonym(s):

D-erythro-Sphingosine (C14 Base)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$417.00

$417.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$417.00

About This Item

Empirical Formula (Hill Notation):
C14H29NO2
CAS Number:
Molecular Weight:
243.39
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$417.00


Estimated to ship on28 May 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (860670P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860670P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](N)([H])[C@]([H])(O)/C=C/CCCCCCCCC

InChI

1S/C14H29NO2/c1-2-3-4-5-6-7-8-9-10-11-14(17)13(15)12-16/h10-11,13-14,16-17H,2-9,12,15H2,1H3/b11-10+/t13-,14+/m0/s1

InChI key

VDRZDTXJMRRVMF-NXFSIWHZSA-N

General description

Sphingosine (d14:1), also referred as tetradecasphing-4-enine, is a C14 analog of sphingosine. It is a major part of sphingoid bases in Musca domestica larvae and adults.[1]

Biochem/physiol Actions

Sphingosine acts as a negative regulator of cell proliferation. It induces apoptosis.[2] It modulates the activities of enzymes such as, phospholipases and protein kinases. In addition, sphingosine also regulate ion channels, cannabinoid receptor type 1 (CB-1) receptors and steroidogenic factor 1 (SF-1) receptors.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (860670P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

the
Lipid Mediators and Their Metabolism in the Brain (2011)
Olivier Cuvillier
Biochimica et biophysica acta, 1585(2-3), 153-162 (2003-01-18)
The sphingolipid metabolites ceramide, sphingosine, and sphingosine 1-phosphate contribute to controlling cell proliferation and apoptosis. Ceramide and its catabolite sphingosine act as negative regulators of cell proliferation and promote apoptosis. Conversely, sphingosine 1-phosphate, formed by phosphorylation of sphingosine by a
The occurrence of tetradecasphing-4-enine and hexadecasphing-4-enine as the principal sphingosines of Musca domestica larvae and adults.
L L Bieber et al.
Biochimica et biophysica acta, 187(1), 157-159 (1969-07-29)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service