Skip to Content
Merck
All Photos(2)

Key Documents

855276P

Avanti

09:0 Lyso PC

1-nonanoyl-2-hydroxy-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-nonanoyl-sn-glycero-3-phosphocholine; PC(9:0/0:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$328.00

$328.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$328.00

About This Item

Empirical Formula (Hill Notation):
C17H36NO7P
CAS Number:
Molecular Weight:
397.44
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$328.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 25 mg (855276P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855276P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCCC)=O

InChI key

VKTRCGYRHHVHRW-MRXNPFEDSA-N

General description

09:0 Lyso PC is a short-chain lysophosphatidylcholine (lysoPC). It is polar in nature.[1]

Application

09:0 Lyso PC may be used in thin-layer chromatography (TLC) analysis for lipid extraction.[1]

Packaging

5 mL Clear Glass Sealed Ampule (855276P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preferential hydrolysis of truncated oxidized glycerophospholipids by lysosomal phospholipase A2
Abe A, et al.
Journal of Lipid Research, 58(2), 339-349 (2017)
R E Stafford et al.
Biochemistry, 28(12), 5113-5120 (1989-06-13)
The critical micelle concentration (cmc) of several lysophospholipids and of a lysophospholipid analogue was determined from surface tension measurements using the maximum bubble pressure method and/or 31P NMR. The use of the maximum bubble pressure method has now been extended

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service