Skip to Content
Merck
All Photos(1)

Key Documents

171204P

Avanti

Heart Extract Polar

Avanti Research - A Croda Brand

Synonym(s):

Heart Polar Lipid Extract (Bovine)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$373.00

$373.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$373.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$373.00


Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 100 mg (171204P-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

lipid extracts

shipped in

dry ice

storage temp.

−20°C

General description

Heart Extract Polar comprises the neutral lipid as a major component. Other components include phosphatidylcholine, phosphatidylserine, phosphatidylethanolamine and phosphatidylinositol.
Polar Lipid Extract is derived from the total lipid extract by precipitation with acetone followed by extraction of the acetone insoluble material with diethyl ether.

Application

Heart Extract Polar may be used:
  • in the preparation of lipid bilayer membrane for ion channel ICln reconstitution[1]
  • in preparation of model membranes for flow fluorometry studies[2]
  • as a lipid standard in thin-layer chromatography[3]

Biochem/physiol Actions

The lipids from the bovine heart mitochondria display a higher phospholipid content compared to liver lipids.[4] Lipid extracts from the mammalian tissues like heart, kidney and liver are used in generating model membranes for mimicking real membranes. These lipid extracts also display nonlamellar phases majorly pertaining to hexagonal, cubic, micellar types at physiological temperatures.[2]

Packaging

5 mL Clear Glass Sealed Ampule (171204P-100mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ICln channels reconstituted in heart-lipid bilayer are selective to chloride
Garavaglia M, et al.
Pflugers Archiv : European Journal of Physiology, 443(5-6), 748-753 (2002)
Lipid composition of mitochondria from bovine heart, liver, and kidney
Fleischer S, et al.
Journal of Lipid Research, 8(3), 170-180 (1967)
Inorganic mercury and cadmium induce rigidity in eukaryotic lipid extracts while mercury also ruptures red blood cells
Kerek E, et al.
Biochimica et Biophysica Acta - Biomembranes, 1860(3), 710-717 (2018)
Natural lipid extracts and biomembrane-mimicking lipid compositions are disposed to form nonlamellar phases, and they release DNA from lipoplexes most efficiently
Koynova R and MacDonald RC
Biochimica et Biophysica Acta - Biomembranes, 1768(10), 2373-2382 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service