Skip to Content
Merck
All Photos(1)

Key Documents

852479L

Avanti

C18(Plasm) LPC-d9

Avanti Research - A Croda Brand

Synonym(s):

1-O-1′-(Z)-octadecenyl(d9)-2-hydroxy-sn-glycero-3-phosphocholine, Plasmalogen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H45D9NO6P
Molecular Weight:
516.74
UNSPSC Code:
12352211
NACRES:
NA.21
Product 852479L is not currently sold in your country. Contact Technical Service

Assay

>99% (TLC)

form

(DCM:Methanol (1:1) solution)

packaging

pkg of 1 × 5 mg (852479L-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics

shipped in

dry ice

storage temp.

−20°C

Application

Lipidomics

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service