Skip to Content
Merck
All Photos(2)

Key Documents

850095P

Avanti

Brain Lyso PE

Avanti Research - A Croda Brand 850095P, powder

Synonym(s):

L-α-lysophosphatidylethanolamine plasmalogen (Brain, Porcine)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$616.00

$616.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$616.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

$616.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (LPE; may contain up to 10% of the 2-LPE isomer, TLC)

form

powder

packaging

pkg of 1 × 10 mg (850095P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850095P

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC/C=C\OC[C@](O)([H])COP([O-])(OCC[NH3+])=O

InChI

1S/C23H48NO6P/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-28-21-23(25)22-30-31(26,27)29-20-18-24/h17,19,23,25H,2-16,18,20-22,24H2,1H3,(H,26,27)/b19-17-/t23-/m1/s1

InChI key

KACDZDULGKPXHT-HIVNOOBXSA-N

General description

Lysophosphotidylethanolamine (LPE) is a phospholipid that belongs to a group called as cephalins.[1] It is a membrane glycerolipid.[2]

Application

Brain Lyso PE has been used as a calibrant in ion mobility-mass spectrometry (IM-MS). It has also been used as a calibrant in hydrophilic interaction chromatography.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (850095P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysophosphatidylethanolamine effects on horticultural commodities: A review
Amaro, Ana L and Almeida, Domingos PF
Postharvest biology and technology, 78(1), 92-102 (2013)
Phosphatidylethanolamine and lysophosphatidylethanolamine
Robins, DC
The Journal of Pharmacy and Pharmacology, 15(1), 701-722 (1963)
Marianne S Jurkowitz et al.
Biochimica et biophysica acta, 1848(2), 742-751 (2014-12-03)
Lysoplasmalogenase catalyzes hydrolytic cleavage of the vinyl-ether bond of lysoplasmalogen to yield fatty aldehyde and glycerophospho-ethanolamine or glycerophospho-choline. We recently purified lysoplasmalogenase from rat liver microsomes and identified the protein as TMEM86B, an integral membrane protein that is a member
Assessment of altered lipid homeostasis by HILIC-ion mobility-mass spectrometry-based lipidomics
Hines K M, et al.
Journal of Lipid Research, 58(4), 809-819 (2017)
Ieva Ailte et al.
Scientific reports, 6, 30336-30336 (2016-07-28)
Shiga toxin (Stx), an AB5 toxin, binds specifically to the neutral glycosphingolipid Gb3 at the cell surface before being transported into cells. We here demonstrate that addition of conical lysophospholipids (LPLs) with large head groups inhibit Stx binding to cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service