Skip to Content
Merck
All Photos(2)

Key Documents

850543P

Avanti

8.9 MAG

1-(8Z-heptadecenoyl)-rac-glycerol, powder

Synonym(s):

17:1 MG (8.9)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$377.00

$377.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$377.00

About This Item

Empirical Formula (Hill Notation):
C20H38O4
CAS Number:
Molecular Weight:
342.51
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$377.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

mol wt

342.51 g/mol

packaging

pkg of 1 × 25 mg (with screw cap (850543P-25mg))

manufacturer/tradename

Avanti Research - A Croda Brand 850543P

technique(s)

protein sequencing: suitable

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=C(CCCCCC/C=C\CCCCCCCC)OCC(O)CO

InChI

1S/C20H38O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-20(23)24-18-19(22)17-21/h9-10,19,21-22H,2-8,11-18H2,1H3/b10-9-

InChI key

UQZGIPIXMWOKEC-KTKRTIGZSA-N

General description

Monoacylglycerols (MAGs) are glycerol esters.[1]
This new Lipid detergent may be used for crystallizing membrane proteins by using lipidic mesophases. This method has variously been referred to as the lipidic cubic phase or in meso method. The method has been shown to be quite versatile in that it has been used to solve X-ray crystallographic structures of prokaryotic and eukaryotic proteins, proteins that are monomeric, homo- and hetero-multimeric, chromophore-containing and chromophore-free, and alpha-helical and beta-barrel proteins.

Biochem/physiol Actions

Monoacylglycerols (MAGs) are usually preferred in food industry as emulsifiers.[1] They are used in several cosmetics products. It is also useful in plastic and textile industry.[1]

Packaging

5 mL PTFE Vial with Screw Cap (850543P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of monoacylglycerols through glycerolysis of ethyl esters derived from linseed oil by green processes
Hobuss CB, et al.
Royal Society of Chemistry Advances, 10(4), 2327-2336 null
Niklaus Johner et al.
Journal of the American Chemical Society, 136(8), 3271-3284 (2014-02-06)
The recent advances in the in meso crystallization technique for the structural characterization of G-protein coupled receptor (GPCR) proteins have established the usefulness of the lipidic-cubic phases (LCPs) in the field of crystallography of membrane proteins. It is surprising that
Martin Caffrey et al.
Journal of visualized experiments : JoVE, (45) (2010-11-30)
A detailed protocol for crystallizing membrane proteins by using lipidic mesophases is described. This method has variously been referred to as the lipidic cubic phase or in meso method. The method has been shown to be quite versatile in that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service