Skip to Content
Merck
All Photos(1)

Key Documents

1.08803

Supelco

Drum key

for use with containers with 2" and 3/4" screw caps

Synonym(s):

Drum key, Barrel key

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
$195.00

$195.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 UNIT
$195.00

About This Item

UNSPSC Code:
41122409
NACRES:
NB.51

$195.00


Estimated to ship on29 May 2025


Request a Bulk Order

compatibility

for use with containers with 2" and 3/4" screw caps

storage temp.

no temp limit

General description

The specially designed drum key is used to open our barrels and drums with 2 and 3/4" opening.
for opening and closing containers with 2" and 3/4" screw caps

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service