Skip to Content
Merck
All Photos(2)

Key Documents

850511P

Avanti

n-octyl-β-D-glucoside

Avanti Research - A Croda Brand

Synonym(s):

octyl glucoside; OG; OGP

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$274.00
5 G
$1,060.00

$274.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$274.00
5 G
$1,060.00

About This Item

Empirical Formula (Hill Notation):
C14H28O6
CAS Number:
Molecular Weight:
292.37
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25

$274.00


Check Cart for Availability

Request a Bulk Order

description

n-octyl-β-D-glucopyranoside

Assay

>99% (contains <0.2% alpha isomer, TLC)

form

powder

mol wt

292.37 g/mol

packaging

pkg of 1 × 1 g (850511P-1g)
pkg of 1 × 5 g (850511P-5g)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](OCCCCCCCC)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C14H28O6/c1-2-3-4-5-6-7-8-19-14-13(18)12(17)11(16)10(9-15)20-14/h10-18H,2-9H2,1H3/t10-,11-,12+,13-,14-/m1/s1

InChI key

HEGSGKPQLMEBJL-RKQHYHRCSA-N

General description

n-octyl-β-D-glucoside (βOG) is a non-ionic detergent.[1] It has a eight carbon hydrophobic tail.[2]
Octyl glucoside is a dialyzable nonionic detergent for functional solubilization and purification of membrane proteins.

Application

n-octyl-β-D-glucoside is suitable for use to study its self-diffusion coefficients in α-cyclodextrin and β-cyclodextrin solutions at 25°C. [3]

Biochem/physiol Actions

n-octyl-β-D-glucoside (βOG) helps to extract, solubilize and denature membrane proteins. Octyl glucoside has versatile uses including: reconstituting an ion channel, solubilizing an enzyme and incorporating a protein into liposomes. Also, nonionizing detergent complexes are used in protein crystallization studies. It is mainly used in the expression and purification of recombinant protein.[2]

Packaging

20 mL Clear Glass Screw Cap Vial (850511P-1g)
30 mL Amber Wide Mouth Glass Bottle with Screw Cap (850511P-5g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A single beta-octyl glucoside molecule induces HIV-1 Nef dimer formation in the absence of partner protein binding
Wu M, et al.
Testing, 13(2), e0192512-e0192512 (2018)
APOBEC3G is incorporated into virus-like particles by a direct interaction with HIV-1 Gag nucleocapsid protein
Alce TM and Popik W
The Journal of Biological Chemistry, 279(33), 34083-34086 (2004)
Interactions between n-octyl and n-nonyl beta-D-glucosides and alpha-and beta-cyclodextrins as seen by self-diffusion NMR
Valente AJM, et al.
Journal of Colloid and Interface Science, 281(1), 218-224 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service