Skip to Content
Merck
All Photos(2)

Key Documents

850500P

Avanti

CHAPS

Avanti Research - A Croda Brand

Synonym(s):

3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$102.00

$102.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$102.00

About This Item

Empirical Formula (Hill Notation):
C32H58N2O7S
CAS Number:
Molecular Weight:
614.88
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.25

$102.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

mol wt

614.88 g/mol

packaging

pkg of 1 × 1 g (850500P-1g)

manufacturer/tradename

Avanti Research - A Croda Brand

technique(s)

protein quantification: suitable

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@@]12C(CC[C@@H]2[C@@H](CCC(NCCC[N+](C)(CCCS(=O)([O-])=O)C)=O)C)C3[C@H](O)C[C@]4([H])C[C@H](O)CC[C@]4(C)C3C[C@@H]1O

InChI

1S/C32H58N2O7S/c1-21(8-11-29(38)33-14-6-15-34(4,5)16-7-17-42(39,40)41)24-9-10-25-30-26(20-28(37)32(24,25)3)31(2)13-12-23(35)18-22(31)19-27(30)36/h21-28,30,35-37H,6-20H2,1-5H3,(H-,33,38,39,40,41)/t21-,22+,23-,24-,25+,26+,27-,28+,30+,31+,32-/m1/s1

InChI key

UMCMPZBLKLEWAF-BCTGSCMUSA-N

General description

CHAPS is a nondenaturing zwitterionic detergent for membrane biochemistry. Useful for solubilizing membrane proteins and breaking protein-protein interactions. CHAPS′ small micellar molecular weight (6,150; aggregation number = 10) and high critical micelle concentration (6-10 mM) allow it to be removed from samples by dialysis. It is also suitable for protein solubilization for isoelectric focusing and two-dimensional electrophoresis. CHAPS is commonly used for non-denaturing (without urea) IEF and has been shown to give excellent resolution of some subcellular preparations and plant proteins. Concentrations between 2-4% (w/v) are typically used in an IEF gel.

Application

CHAPS is suitable to prepare bicelles[1] and to study the processes of liposome solubilization and reconstitution of transport proteins.[2]

Packaging

20 mL Clear Glass Screw Cap Vial (850500P-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Bilayer membrane modulation of membrane type 1 matrix metalloproteinase (MT1-MMP) structure and proteolytic activity
Cerofolini L, et al.
Scientific reports, 6, 29511-29511 (2016)
Liposome solubilization and membrane protein reconstitution using Chaps and Chapso
Cladera J, et al.
European Journal of Biochemistry, 243(3), 798-804 (1997)
Functional reconstitution of the nicotinic acetylcholine receptor by CHAPS dialysis depends on the concentrations of salt, lipid, and protein
Schuerholz T, et al.
Biochemistry, 31(21), 5067-5077 (1992)
Stuart J Cordwell
Methods in molecular biology (Clifton, N.J.), 424, 139-146 (2008-03-29)
Reproducible techniques for the prefractionation of proteins prior to two-dimensional gel electrophoresis (2-DE) are essential for increasing the number of unique proteins that can be identified and assayed following biological experimentation. A simple and robust technique for separating highly soluble
Erik F Kot et al.
Langmuir : the ACS journal of surfaces and colloids, 34(11), 3426-3437 (2018-02-28)
Isotropic phospholipid bicelles are one of the most prospective membrane mimetics for the structural studies of membrane proteins in solution. Recent works provided an almost full set of data regarding the properties of isotropic bicelles; however, one major aspect of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service