Skip to Content
Merck
All Photos(2)

Key Documents

850464C

Avanti

18:0-14:0 PC

1-stearoyl-2-myristoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-octadecanoyl-2-tetradecanoyl-sn-glycero-3-phosphocholine; SMPC; PC(18:0/14:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$245.00

$245.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$245.00

About This Item

Empirical Formula (Hill Notation):
C40H80NO8P
CAS Number:
Molecular Weight:
734.04
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$245.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850464C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850464C

concentration

10 mg/mL (850464C-25mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylcholine (PC) is a major bilayer-forming phospholipid in which the headgroup carries a positively and negatively charged quaternary amine and a phosphate, respectively.[1]

Application

18:0-14:0 PC (1-stearoyl-2-myristoyl-sn-glycero-3-phosphocholine) may be used in liposomes to study its interference in the proliferation of several cell lines using transmission electron microscopy (TEM).[2]

Biochem/physiol Actions

Phosphatidylcholine (PC) can form a hydrophobic surface in the mucus by acting as a surfactant to inhibit the penetrance of bacteria.[3]

Packaging

5 mL Clear Glass Sealed Ampule (850464C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipid-Protein Interactions in Membranes
The Membranes of Cells, 291-334 (2016)
Inflammatory Bowel Disease
Integrative Medicine, 501-516 (2018)
Anna Werlein et al.
Biochimica et biophysica acta, 1848(7), 1599-1608 (2015-04-29)
According to early experiments with natural extracts, phosphatidylcholines (PCs) are widely considered essentially non-toxic. In addition to these physiological mixed-chain PCs, many different synthetic diacyl-PCs are currently available, but they have never been systematically evaluated for any interference with cell
Simon Becher et al.
Analytical chemistry, 90(19), 11486-11494 (2018-09-11)
Phosphatidylcholines are the major phospholipid component of most eukaryotic cell membranes. Phosphatidylcholines have been shown to actively participate in regulatory and metabolic processes. Dysfunctional metabolic processes have been linked to human disease and can result in altered phosphatidylcholine structural features

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service