Skip to Content
Merck
All Photos(2)

Key Documents

850368P

Avanti

20:0 PC

1,2-diarachidoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1,2-dieicosanoyl-sn-glycero-3-phosphocholine; PC(20:0/20:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$209.00
200 MG
$377.00
500 MG
$599.00

$209.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$209.00
200 MG
$377.00
500 MG
$599.00

About This Item

Empirical Formula (Hill Notation):
C48H96NO8P
CAS Number:
Molecular Weight:
846.25
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$209.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (850368P-200mg)
pkg of 1 × 25 mg (850368P-25mg)
pkg of 1 × 500 mg (850368P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850368P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCCCC)=O)=O

InChI

1S/C48H96NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-38-40-47(50)54-44-46(45-56-58(52,53)55-43-42-49(3,4)5)57-48(51)41-39-37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h46H,6-45H2,1-5H3/t46-/m1/s1

InChI key

YKIOPDIXYAUOFN-YACUFSJGSA-N

General description

Phosphatidylcholine, a major source of diacylglycerol (DAG) is considered as a biological membrane lipid.[1] 20:0 PC is a zwitterionic lipid.[2]
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

20:0 PC may be used:
  • in the preparation of liposomes[2]
  • to prepare vesicles or supported bilayer to study its ability in spontaneously forming distinct lipid domains, rafts and/or asymmetric bilayers along with other phosphatidylcholine lipids[3]
  • to prepare small unilamellar vesicles (SUVs) to study the effect of hydrophobic mismatch on gramicidin conformation and organization[4]

Biochem/physiol Actions

Phosphatidylcholine (PC) can serve as a surfactant in the mucus.[5]

Packaging

20 mL Clear Glass Screw Cap Vial (850368P-500mg)
5 mL Amber Glass Screw Cap Vial (850368P-200mg)
5 mL Amber Glass Screw Cap Vial (850368P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Second Messengers
Cell Biology, 443-462 (2017)
Inflammatory Bowel Disease
Integrative Medicine, 501-516 (2018)
Markus Ritter et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 32(1), 46-52 (2013-07-23)
We investigated if mixtures of the phosphatidylcholine (PC) lipids 1,2-dilauroyl-sn-glycero-3-phosphocholine (C12:0 PC; DLPC) and 1,2-diarachidoyl-sn-glycero-3-phosphocholine (C20:0 PC; DAPC), which differ by eight methylene groups in acyl chain length, lead to the spontaneous formation of distinct lipid rafts and asymmetric bilayers.
Devaki A Kelkar et al.
Biochimica et biophysica acta, 1768(5), 1103-1113 (2007-02-27)
The matching of hydrophobic lengths of integral membrane proteins and the surrounding lipid bilayer is an important factor that influences both structure and function of integral membrane proteins. The ion channel gramicidin is known to be uniquely sensitive to membrane
Federico Battisti et al.
Frontiers in immunology, 8, 1179-1179 (2017-10-11)
Dendritic cells (DCs) are the only antigen-presenting cells able to prime naïve T cells and cross-prime antigen-specific CD8+ T cells. Their functionality is a requirement for the induction and maintenance of long-lasting cancer immunity. Albeit intensively investigated, the in vivo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service