Skip to Content
Merck
All Photos(2)

Key Documents

840862P

Avanti

18:0-18:2 PA

1-stearoyl-2-linoleoyl-sn-glycero-3-phosphate (sodium salt), powder

Synonym(s):

1-octadecanoyl-2-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phosphate (sodium salt); PA(18:0/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H72O8PNa
CAS Number:
Molecular Weight:
722.95
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (840862P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840862P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C39H73O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(40)45-35-37(36-46-48(42,43)44)47-39(41)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h12,14,18,20,37H,3-11,13,15-17,19,21-36H2,1-2H3,(H2,42,43,44);/q;+1/p-1/b14-12-,20-18-;/t37-;/m1./s1

InChI key

QCCKEVYCMGRONT-ZNKFQGFOSA-M

General description

Phosphatidic acid (PA) is an amphipathic lipid, containing a polar head group and two long hydrophobic acyl chains. PA is anionic in nature and a membrane structural lipid. It acts as a parent compound for more complex membrane phospholipids.

Biochem/physiol Actions

Phosphatidic acid (PA) is a bioactive lipid and a secondary messenger. PA reacts with water and other phospholipids to produce lipid bilayers and multicomponent bilayers respectively.

Packaging

5 mL Clear Glass Sealed Ampule (840862P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipid second messengers and Receptors
Encyclopedia of Endocrine Diseases, 77(3), 329-334 (2004)
Bioactive Lipids
An Introduction to Biological Membranes, 453-478 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service