Skip to Content
Merck
All Photos(2)

Key Documents

840635P

Avanti

12:0 PA

1,2-dilauroyl-sn-glycero-3-phosphate (sodium salt), powder

Synonym(s):

1,2-didodecanoyl-sn-glycero-3-phosphate (sodium salt); DLPA; PA(12:0/12:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$195.00
200 MG
$577.00

$195.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$195.00
200 MG
$577.00

About This Item

Empirical Formula (Hill Notation):
C27H52O8PNa
CAS Number:
Molecular Weight:
558.66
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$195.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (840635P-200mg)
pkg of 1 × 25 mg (840635P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840635P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O)=O)(OC(CCCCCCCCCCC)=O)COC(CCCCCCCCCCC)=O.[Na+]

InChI

1S/C27H53O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-26(28)33-23-25(24-34-36(30,31)32)35-27(29)22-20-18-16-14-12-10-8-6-4-2;/h25H,3-24H2,1-2H3,(H2,30,31,32);/q;+1/p-1/t25-;/m1./s1

InChI key

CIOHBYXGWWPOIS-VQIWEWKSSA-M

General description

12:0 PA (1,2-dilauroyl-sn-glycero-3-phosphate) is a sodium salt of modified lipid, wherein C-1, C-2 of glycerol is substituted with didodecanoyl and C-3 of glycerol is substituted with phosphatidic acid group.

Application

12:0 PA (1,2-dilauroyl-sn-glycero-3-phosphate (sodium salt)) has been used as an internal lipid standard in the matrix-assisted laser desorption/ionization Fourier transform ion cyclotron resonance mass spectrometry (MALDI FT-ICR MS) analysis.[1] It may be used as a lipid standard:
  • for the quantification of phospholipids by high-performance liquid chromatography coupled to tandem mass spectrometryelectrospray ionization tandem mass spectrometry (HPLC/ESI-MS/MS)[2]
  • for the quantification of lipids in cold-pressed rapeseed oils based on direct infusion electrospray ionization tandem mass spectrometry[3]
  • in liquid chromatography tandem mass spectrometry (LC-MS/MS) analysis[4]

Biochem/physiol Actions

Phosphatidic acid (PA) might serve as a modulator of phagocyte respiratory burst.[5]

Packaging

5 mL Amber Glass Screw Cap Vial (840635P-200mg)
5 mL Amber Glass Screw Cap Vial (840635P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie Garnier-Lhomme et al.
PloS one, 4(1), e4255-e4255 (2009-01-24)
The cytoplasm of eukaryotic cells is a highly dynamic compartment where membranes readily undergo fission and fusion to reorganize the cytoplasmic architecture, and to import, export and transport various cargos within the cell. The double membrane of the nuclear envelope
Glycerolipids in signal transduction
New comprehensive biochemistry, 31, 237-255 null
Richard D Byrne et al.
Cellular signalling, 19(5), 913-922 (2006-12-23)
Nuclear envelope assembly is an essential event in each cell cycle but the proteins and lipids involved in its regulation remain mostly unknown. Assembly involves membrane fusions but neither specific SNAREs nor Rab GTPases have been identified in its control.
Ya Xie et al.
Food chemistry, 285, 194-203 (2019-02-25)
Microwave pretreatment of rapeseeds prior to cold-pressing is a simple and desirable method for producing high quality oils. In this study, a rapid and sensitive lipid profiling platform employing an accurate quantification strategy was established based on direct infusion electrospray
Yuchen Sun et al.
Metabolites, 9(11) (2019-11-07)
Extracellular vesicles (EVs) consist of lipid bilayers, occur in various biofluids, and are invaluable in biomarker screening. Liquid chromatography coupled with high-resolution mass spectrometry (LC-MS) was recently used to study comprehensive EV lipid profiles in vitro. The aim of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service