Skip to Content
Merck
All Photos(2)

Key Documents

830855P

Avanti

16:0 PA

Avanti Research - A Croda Brand

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphate (sodium salt); DPPA; PA(16:0/16:0); 110614

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$136.00
200 MG
$471.00
500 MG
$866.00

$136.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$136.00
200 MG
$471.00
500 MG
$866.00

About This Item

Empirical Formula (Hill Notation):
C35H68O8PNa
CAS Number:
Molecular Weight:
670.87
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$136.00


Check Cart for Availability

Request a Bulk Order

description

1,2-dipalmitoyl-sn-glycero-3-phosphate (sodium salt)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 200 mg (830855P-200mg)
pkg of 1 × 25 mg (830855P-25mg)
pkg of 1 × 500 mg (830855P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[Na+].[P](=O)([O-])([O-])OC[C@H](OC(=O)CCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCC

InChI

1S/C35H69O8P.2Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(36)41-31-33(32-42-44(38,39)40)43-35(37)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;;/h33H,3-32H2,1-2H3,(H2,38,39,40);;/q;2*+1/p-2/t33-;;/m1../s1

InChI key

NFRFUGBXJTXTMZ-QYKZUBHNSA-L

General description

Phosphatidic acid is obtained from glycerophospholipid with the help of phospholipase D.[1]

Application

16:0 PA may be used as lipid standards for the quantification of the lipid species released from E-SYT2 expressed in mammalian cells.[2] It may also be used in the synthesis of the methylene blue microbubbles.[3]

Biochem/physiol Actions

Phosphatidic acid might serve as a modulator of phagocyte respiratory burst. It also acts as a precursor of diacylglycerol and lysophosphatidic acid.[1] 16:0 PA/DPPA can improve the bactericidal effect of eugenol.[4]

Packaging

20 mL Clear Glass Screw Cap Vial (830855P-500mg)
5 mL Amber Glass Screw Cap Vial (830855P-200mg)
5 mL Amber Glass Screw Cap Vial (830855P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lorissa J Smulan et al.
Cell reports, 16(1), 9-18 (2016-06-21)
Lipogenesis requires coordinated expression of genes for fatty acid, phospholipid, and triglyceride synthesis. Transcription factors, such as SREBP-1 (Sterol regulatory element binding protein), may be activated in response to feedback mechanisms linking gene activation to levels of metabolites in the
Methylene blue microbubbles as a model dual-modality contrast agent for ultrasound and activatable photoacoustic imaging
Jeon M, et al.
Journal of Biomedical Optics, 19(1), 016005-016005 (2014)
Critical synergistic concentration of lecithin phospholipids improves the antimicrobial activity of eugenol against Escherichia coli
Zhang H, et al.
Applied and Environmental Microbiology, 83(21), e01583-e01517 (2017)
Structure of a lipid-bound extended synaptotagmin indicates a role in lipid transfer
Schauder CM, et al.
Nature, 510(7506), 552-552 (2014)
Angelina Provost et al.
Scientific reports, 9(1), 17967-17967 (2019-12-01)
Direct stochastic optical reconstruction microscopy (dSTORM), developed in the last decade, has revolutionised optical microscopy by enabling scientists to visualise objects beyond the resolution provided by conventional microscopy (200 nm). We developed an innovative method based on blinking particle standards and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service