Skip to Content
Merck
All Photos(3)

Key Documents

840524P

Avanti

DGDG

Avanti Research - A Croda Brand

Synonym(s):

1,2-diacyl-3-O-(α-D-galactosyl1-6)-α-D-galactosyl-sn-glycerol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$377.00
10 MG
$655.00
25 MG
$1,390.00

$377.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$377.00
10 MG
$655.00
25 MG
$1,390.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

$377.00


Check Cart for Availability

Request a Bulk Order

description

Digalactosyldiacylglycerol (Plant)

Assay

>99% (TLC)

form

powder

mol wt

~926.77 g/mol (average based on fatty acid distribution)

packaging

pkg of 1 × 5 mg (840524P-5mg)
pkg of 2 × 5 mg (840524P-10mg)
pkg of 5 × 5 mg (840524P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phosphoglycerides
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](COC(CCCCCCC/C=C\C/C=C\C/C=C\CC)=O)(OC(CCCCCCC/C=C\C/C=C\C/C=C\CC)=O)CO[C@H](O1)[C@H](O)[C@@H](O)[C@H]([C@H]1CO[C@H](O2)[C@H](O)[C@@H](O)[C@H]([C@H]2CO)O)O

InChI key

KDYAPQVYJXUQNY-KEOSPFNWSA-N

General description

Digalactosyldiacylglycerol (DGDG) is vital for photosynthesis.[1]
Monogalactosyldiacylglycerol (MGDG) and digalactosyldiacylglycerol (DGDG) are the two nonionic lipid constituents of the thylakoid membrane of higher plant chloroplasts. MGDG and DGDG are present in the membrane at 56% and 29%, respectively, of the total lipid content. DGDG is a bilayer-forming lipid, while MGDG alone will only form hexagonal-II structures.

Application

Digalactosyldiacylglycerol (DGDG) has been used in hydrophilic interaction LC-ion mobility-MS (HILIC-IM-MS) for untargeted lipidomics.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (840524P-10mg)
5 mL Amber Glass Screw Cap Vial (840524P-25mg)
5 mL Amber Glass Screw Cap Vial (840524P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reduced biosynthesis of digalactosyldiacylglycerol, a major chloroplast membrane lipid, leads to oxylipin overproduction and phloem cap lignification in Arabidopsis
Lin YT, et al.
Plant Cell, 28(1), 219-232 (2016)
Weihua Ye et al.
The journal of physical chemistry. B, 117(4), 1044-1050 (2013-01-11)
Galactolipids are the main structural component of plant chloroplastic (thylakoid) membranes and of blue-green algae cell membranes. The predominant lipids in this class are monogalactosyl-diacylglycerol (MGDG) and digalactosyl-diacylglycerol (DGDG). We here present a method for the preparation of bicelles that
Kuniyoshi Kaseda et al.
Pharmacological research, 152, 104633-104633 (2020-01-10)
Advanced glycation end products (AGEs) and their receptor (RAGE) system evoke inflammatory reactions and insulin resistance in adipocytes. Spa-derived green alga Mucidosphaerium sp. (MS) had anti-inflammatory properties in vitro. We examined here whether and how MS could ameliorate insulin resistance
Characterization of the mechanisms of daptomycin resistance among Gram-positive bacterial pathogens by multidimensional lipidomics
Hines KM, et al.
mSphere, 2(6), e00492-e00417 (2017)
Cyrille Y Botté et al.
The Journal of biological chemistry, 286(34), 29893-29903 (2011-06-30)
Apicomplexa are protist parasites that include Plasmodium spp., the causative agents of malaria, and Toxoplasma gondii, responsible for toxoplasmosis. Most Apicomplexa possess a relict plastid, the apicoplast, which was acquired by secondary endosymbiosis of a red alga. Despite being nonphotosynthetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service