Skip to Content
Merck
All Photos(3)

Key Documents

840022P

Avanti

Brain PE

Avanti Research - A Croda Brand 840022P, powder

Synonym(s):

L-α-phosphatidylethanolamine (Brain, Porcine)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$978.00
200 MG
$2,860.00

$978.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$978.00
200 MG
$2,860.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

$978.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 2 × 100 mg (840022P-200mg)
pkg of 1 × 25 mg (840022P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840022P

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O

InChI

1S/C41H80NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-40(43)47-37-39(38-49-51(45,46)48-36-35-42)50-41(44)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h18,20,39H,3-17,19,21-38,42H2,1-2H3,(H,45,46)/b20-18-/t39-/m1/s1

InChI key

JQKOHRZNEOQNJE-DJEJVYNPSA-N

General description

Contains ~50% plasmalogen ether lipid

Application

Brain PE has been used for the preparation of liposomes.[1][2]

Packaging

5 mL Clear Glass Sealed Ampule (840022P-200mg)
5 mL Clear Glass Sealed Ampule (840022P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

RIBEYE (B)-domain binds to lipid components of synaptic vesicles in an NAD (H)-dependent, redox-sensitive manner
Schwarz K and Schmitz F
The Biochemical Journal, 474(7), 1205-1220 (2017)
Saurabh Srivastava et al.
PloS one, 10(6), e0130283-e0130283 (2015-06-20)
Recent studies introduced two experimental protocols for converting full-length recombinant prion protein (rPrP) purified from E.coli into the infectious prion state (PrPSc) with high infectivity titers. Both protocols employed protein misfolding cyclic amplification (PMCA) for generating PrPSc de novo, but
Qualitative and quantitative characterization of protein-phosphoinositide interactions with liposome-based methods
Busse RA, et al.
Autophagy, 9(5), 770-777 (2013)
Arupratan Das et al.
Nature cell biology, 14(3), 304-310 (2012-02-22)
Lipid asymmetry at the plasma membrane is essential for such processes as cell polarity, cytokinesis and phagocytosis. Here we find that a lipid flippase complex, composed of Lem3, Dnf1 or Dnf2, has a role in the dynamic recycling of the
Gillian L Dornan et al.
Structure (London, England : 1993), 28(2), 145-156 (2019-12-14)
The class I phosphoinositide 3-kinases (PI3Ks) are key signaling enzymes composed of a heterodimer of a p110 catalytic subunit and a p85 regulatory subunit, with PI3K mutations being causative of multiple human diseases including cancer, primary immunodeficiencies, and developmental disorders.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service