Skip to Content
Merck
All Photos(1)

Documents

810318P

Avanti

TopFluor-oxPAPC

new

Avanti Research - A Croda Brand

Synonym(s):

Oxidized 1-(dipyrrometheneboron difluoride)undecanoyl-2-arachidonoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.79

biological source

synthetic

Assay

>99% (TLC)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC1=CC(C)=[N+]2C1=C(CCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCC=O)c1c(C)cc(C)n1[B-]2(F)F.CC1=CC(C)=[N+]2C1=C(CCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCC(O)=O)c1c(C)cc(C)n1[B-]2(F)F.CC1=CC(C)=[N+]2C1=C(CCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCC=O)c1c(C)cc(C)n1[B-]2(F)F.CC1=CC(C)=[N+]2C1=C(CCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCC(O)=O)c1c(C)cc(C)n1[B-]2(F)F.CC1=CC(C)=[N+]2C1=C(CCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCC(O)\C=C\C(O)=O)c1c(C)cc(C)n1[B-]2(F)F.CC1=CC(C)=[N+]2C1=C(CCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCC(=O)\C=C\C(O)=O)c1c(C)cc(C)n1[B-]2(F)F.CC1=CC(C)=[N+]2C1=C(CCCCCCCCCCC(=O)OC[C@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCC(O)C1OC1C(O)=O)c1c(C)cc(C)n1[B-]2(F)F.CCCCC\C=C/CC1C(O)CC(=O)\C1=C\C1OC1CCCC(=O)O[C@H](COC(=O)CCCCCCCCCCC1=C2C(C)=CC(C)=[N+]2[B-](F)(F)n2c(C)cc(C)c12)COP([O-])(=O)OCC[N+](C)(C)C

Application

Lipids research

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service