Skip to Content
Merck
All Photos(3)

Key Documents

131303P

Avanti

Cerebrosides (Brain)

Avanti Research - A Croda Brand

Synonym(s):

Total Cerebrosides (Brain, Porcine)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$417.00

$417.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$417.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$417.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (131303P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

General description

Cerebrosides are glycosphingolipids with either glucose or galactose as a polar head group. They comprise the major component of the myelin lipids. Cerebrosides are part of non-neural cells including the epithelial cells associated with skin and the digestive system.[1] Cerebrosides are amphipathic in nature.[2]
The total brain cerebrosides appear as a double spot on thin layer chromatography (chloroform/methanol/water, 65:25:4 v/v). The top spot corresponds to ceresine which contain non-hydroxylated fatty acids, and the bottom spot corresponds to phrenosin which contain hydroxylated fatty acids.This Natural Lipid is a mixture and the structure shown above is only representative of one possible structure present in the product. Refer to chart for average fatty acid distribution.

Application

Cerebrosides (Brain) has been used:
  • in the coating of coverslips for neurite outgrowth assay of retinal ganglion cells[3]
  • in reconstruction of the apical membrane of Madin-Darby canine kidney (MDCK) cells[4]
  • in the preparation of liposomes to mimic the MDCK cell membrane for permeability studies[5]

Biochem/physiol Actions

Cerebrosides regulate a variety of physiological processes like cell growth, survival and multiplication. They coordinate membrane trafficking and inhibit apoptosis. Inborn metabolic disorder, Gaucher′s disease is associated with elevated levels of cerebrosides.[6]. Additionally, cerebrosides provide structural support to the membranes and mediate intracellular communication.[2] Accumulation of galactocerebrosides is associated with tumors. It also serves as a receptor for human immune deficiency virus (HIV) glycoprotein.[6] The porcine cerebroside and ganglioside (CPCG) is used in treating neurotoxicity induced by the drug oxaliplatin in digestive system tumors.[7]

Packaging

5 mL Amber Glass Screw Cap Vial (131303P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effects of compound porcine cerebroside and ganglioside on neurotoxicity caused by oxaliplatin chemotherapy: preliminary results.
Zhang YL, et al.
European Review for Medical and Pharmacological Sciences, 23(5), 5441-5448 (2019)
Water permeability of asymmetric planar lipid bilayers: leaflets of different composition offer independent and additive resistances to permeation
Krylov AV, et al.
The Journal of General Physiology, 118(4), 333-340 (2001)
Direct visualization of the lateral structure of porcine brain cerebrosides/POPC mixtures in presence and absence of cholesterol
Fidorra M, et al.
Biophysical Journal, 97(1), 142-154 (2009)
Reconstituting the barrier properties of a water-tight epithelial membrane by design of leaflet-specific liposomes
Hill WG and Zeidel ML
Test, 275(39), 30176-30185 (2000)
The cerebrosides
Tan RX and Chen JH
Natural Product Reports, 20(5), 509-534 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service