Skip to Content
Merck
All Photos(1)

Key Documents

810259C

Avanti

TopFluor Oleic Acid

Avanti Research - A Croda Brand, chloroform solution

Synonym(s):

N-(4-TopFluor butanoyl)-aminooctadec-9-enoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$853.00

$853.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$853.00

About This Item

Empirical Formula (Hill Notation):
C35H54BF2N3O3
CAS Number:
Molecular Weight:
613.63
UNSPSC Code:
12352211
NACRES:
NA.25

$853.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 1 mg (810259C-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL in chloroform (810259C-1MG)

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC1=CC(C)=[N+]2C1=C(CCCC(NCCCCCCCC/C=C\CCCCCCCC(O)=O)=O)C3=C(C)C=C(C)N3[B-]2(F)F

General description

TopFluor is dipyrrometheneboron difluoride undecanoic acid. It is used for labeling lipids. Fluorescent lipids are widely used in live imaging in cell culture and animal models. TopFluor is a green variant of BODIPY (a fluorescent tag for lipids). TopFluor has two additional methyl groups compared to boron-dipyrromethene (BODIPY) and carbon 1 of its central ring is bonded to the lipid conjugate. It forms a T-shaped molecule in contrast to the linear molecule formed by BODIPY FL- fatty acids FAs).[1]

Packaging

5mL Amber Glass Screw Cap Vial (810259C-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An HPLC-CAD/fluorescence lipidomics platform using fluorescent fatty acids as metabolic tracers
Quinlivan VH, et al.
Journal of Lipid Research, 58(5), 1008-1020 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service