Skip to Content
Merck
All Photos(1)

Key Documents

810226P

Avanti

C6-NBD Lactosyl Ceramide

Avanti Research - A Croda Brand 810226P, powder

Synonym(s):

N-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-D-lactosyl-β1-1′-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H69N5O16
CAS Number:
Molecular Weight:
900.02
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810226P-1MG)
pkg of 1 × 250 μg (810226P-250ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810226P

shipped in

dry ice

storage temp.

−20°C

General description

C6-NBD Lactosyl ceramide is a commercially available fluorescent analog of compound lactosyl ceramide.[1] Lactosyl ceramide is present on neutrophils and macrophages.[2]

Application

C6-NBD Lactosyl Ceramide has been used in:
  • lactosylceramide synthase assay as a fluorescent acceptor substrate identified using fluorescence based detector in HPLC(1)
  • phospholipid labeling and fluorescence-lifetime imaging microscopy (FILM) of cell membranes(6)

Biochem/physiol Actions

Lactosyl ceramide is utilized for synthesis of various glycosphingolipids like oligoglycosylceramides and gangliosides in vertebrates. It is an important molecule involved in signaling cascades leading to phenotypic changes such as adhesion, migration, cell proliferation and angiogenesis.[2][3][4][5]

Packaging

5 mL Amber Glass Screw Cap Vial (810226P-1MG)
5 mL Amber Glass Screw Cap Vial (810226P-250ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasuhiro Hayashi et al.
Analytical biochemistry, 345(2), 181-186 (2005-09-06)
Glucosylceramide synthase (GlcT) and lactosylceramide synthase (GalT) are key enzymes for the synthesis of major glycosphingolipids of vertebrates. In this article, we report a new reliable method to determine GlcT and GalT activities using the fluorescent acceptor substrates C6-4-nitrobenzo-2-oxa-1,3-diazole (NBD)-ceramide
Ana Zarubica et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 23(6), 1775-1785 (2009-01-20)
The ABCA1 transporter orchestrates cellular lipid homeostasis by promoting the release of cholesterol to plasmatic acceptors. The molecular mechanism is, however, unknown. We report here on the biophysical analysis in living HeLa cells of the ABCA1 lipid microenvironment at the
W I Weis et al.
Annual review of biochemistry, 65, 441-473 (1996-01-01)
Lectins are responsible for cell surface sugar recognition in bacteria, animals, and plants. Examples include bacterial toxins; animal receptors that mediate cell-cell interactions, uptake of glycoconjugates, and pathogen neutralization; and plant toxins and mitogens. The structural basis for selective sugar
Subroto Chatterjee et al.
Biochimica et biophysica acta, 1780(3), 370-382 (2007-12-14)
Although lactosylceramide (LacCer) plays a pivotal role in the biosynthesis of nearly all the major glycosphingolipids, its function in regulating cellular function has begun to emerge only recently. Our current opinion is that several physiologically critical molecules such as modified/oxidized
S Hakomori et al.
Journal of biochemistry, 118(6), 1091-1103 (1995-12-01)
Glycosphingolipids (GSLs), cell type-specific markers which change dramatically during ontogenesis and oncogenesis, have been implicated as playing major roles in cellular interactions and control of cell proliferation in multicellular organisms. These functional roles have been partially clarified through two types

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service