Skip to Content
Merck
All Photos(1)

Documents

870012P

Avanti

18:1 MPB PE

Avanti Research - A Croda Brand 870012P, powder

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidophenyl)butyramide] (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H88N2O11PNa
CAS Number:
Molecular Weight:
1007.26
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 2 × 100 mg (870012P-200mg)
pkg of 1 × 25 mg (870012P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870012P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(CCCC(C=C1)=CC=C1N(C(C=C2)=O)C2=O)=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C55H89N2O11P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-36-54(61)65-46-50(68-55(62)37-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)47-67-69(63,64)66-45-44-56-51(58)35-33-34-48-38-40-49(41-39-48)57-52(59)42-43-53(57)60;/h17-20,38-43,50H,3-16,21-37,44-47H2,1-2H3,(H,56,58)(H,63,64);/q;+1/p-1/b19-17-,20-18-;/t50-;/m1./s1

InChI key

IILKDCDPRGFBSN-VCRVZWGWSA-M

General description

18:1 MPB PE is a head group modified functionalized lipid. This lipid is used in the preparation of lipidated DNA anti-handles, which are used to label DNA-origami rings involved in the assembly, arrangement, and remodeling of liposomes.

Application

18:1 MPB PE is suitable for use in the preparation of lipidated DNA anti-handles.

Packaging

5 mL Clear Glass Sealed Ampule (870012P-200mg)
5 mL Clear Glass Sealed Ampule (870012P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self-assembly of size-controlled liposomes on DNA nanotemplates
Yang Y, et al.
Nature Chemistry, 8(5), 476-476 (2016)
Placing and shaping liposomes with reconfigurable DNA nanocages
Zhang Z, et al.
Nature Chemistry, 9(7), 653-653 (2017)
Camilla Skyttner et al.
Langmuir : the ACS journal of surfaces and colloids, 34(22), 6529-6537 (2018-05-15)
Membrane-active peptides that enable the triggered release of liposomal cargo are of great interest for the development of liposome-based drug delivery systems but require peptide-lipid membrane interactions that are highly defined and tunable. To this end, we have explored the
Emeka B Okeke et al.
Biomaterials, 238, 119836-119836 (2020-02-12)
Neutrophil elastase (NE) is a serine protease stored in the azurophilic granules of neutrophils and released into the extracellular milieu during inflammatory response or formation of neutrophil extracellular traps (NETs). Neutrophils release NETs to entrap pathogens by externalizing their cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service