Skip to Content
Merck
All Photos(1)

Key Documents

810209X

Avanti

C6-NBD Ceramide

Avanti Research - A Croda Brand 810209X

Synonym(s):

N-[6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl]-D-erythro-sphingosine:methanol (8:2)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$914.00

$914.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$914.00

About This Item

Empirical Formula (Hill Notation):
C30H49N5O6
CAS Number:
Molecular Weight:
575.74
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$914.00


Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810209X-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810209X

concentration

1 mg/mL (810209X-1mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[N+](=O)([O-])c1c2n[o]nc2c(cc1)NCCCCCC(=O)N[C@H]([C@H](O)\C=C\CCCCCCCCCCCCC)CO

InChI

1S/C30H49N5O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-15-18-27(37)25(23-36)32-28(38)19-16-14-17-22-31-24-20-21-26(35(39)40)30-29(24)33-41-34-30/h15,18,20-21,25,27,31,36-37H,2-14,16-17,19,22-23H2,1H3,(H,32,38)/b18-15+/t25-,27+/m0/s1

InChI key

HZIRBXILQRLFIK-VPZZKNKNSA-N

General description

C6-NBD ceramide is a fluorescent labelled compound, known to be useful for the detection of enzymatic action. C6-NBD ceramide is preferred over radioactive compounds.[1]

Application

C6-NBD Ceramide is suitable as a substrate for sphingomyelin synthase activity assay.[2]

Biochem/physiol Actions

Sphingolipid metabolism yields ceramide as one of the products. The family of ceramide synthases catalyses its synthesis. The ceramides differ in its acyl chain length. Ceramides possess a wide range of functions, associated with apoptosis, cancer inhibition and chemotherapy. Its action is also related to neurodegenerative diseases.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (810209X-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system, Liver,Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Glycosyltransferases and Related Genes, 5-5 (2011)
A role for ceramides, but not sphingomyelins, as antagonists of insulin signaling and mitochondrial metabolism in C2C12 myotubes
Park M, et al.
The Journal of Biological Chemistry, 291(46), 23978-23988 (2016)
Mammalian ceramide synthases
Levy M and Futerman AH
IUBMB Life, 62(5), 347-356 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service