Skip to Content
Merck
All Photos(1)

Documents

07-122

Sigma-Aldrich

Anti-Kalirin Antibody

Upstate®, from rabbit

Synonym(s):

Anti-ARHGEF24, Anti-CHDS5, Anti-DUET, Anti-DUO, Anti-HAPIP, Anti-TRAD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KALRN(8997)

Specificity

Kalirin; does not cross react with Trio

Immunogen

GST fusion protein corresponding to residues 517-976 of rat Kalirin, cleaved with thrombin to remove the GST moiety; referred to as JH2582

Application

Anti-Kalirin Antibody is an antibody against Kalirin for use in IP, WB & IC.
Research Category
Signaling
Research Sub Category
G-proteins

Cytoskeletal Signaling

Quality

routinely evaluated by immunoblot on RIPA lysates from rat brain microsomal protein preparation; long exposure times may be required

Target description

190, 115, 370 and 420kDa isoforms

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide prior to the addition of glycerol to 30%
Format: Purified
Protein A chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R E Mains et al.
The Journal of biological chemistry, 274(5), 2929-2937 (1999-01-23)
The production and regulated secretion of bioactive peptides require a series of lumenal enzymes to convert inactive precursors into bioactive peptides plus several cytosolic proteins to govern granule formation, maturation, translocation, and exocytosis. Peptidylglycine alpha-amidating monooxygenase (PAM), an enzyme essential
Kalirin inhibition of inducible nitric-oxide synthase.
Ratovitski, E A, et al.
The Journal of Biological Chemistry, 274, 993-999 (1999)
P Penzes et al.
The Journal of biological chemistry, 275(9), 6395-6403 (2000-02-29)
Communication between membranes and the actin cytoskeleton is an important aspect of neuronal function. Regulators of actin cytoskeletal dynamics include the Rho-like small GTP-binding proteins and their exchange factors. Kalirin is a brain-specific protein, first identified through its interaction with
The Rho-GEF Kalirin regulates bone mass and the function of osteoblasts and osteoclasts.
Huang, S; Eleniste, PP; Wayakanon, K; Mandela, P; Eipper, BA; Mains, RE; Allen, MR; Bruzzaniti, A
Bone null
Xiaoting Wang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(27), 11012-11022 (2013-07-05)
It is well established that behavioral sensitization to cocaine is accompanied by increased spine density and AMPA receptor (AMPAR) transmission in the nucleus accumbens (NAc), but two major questions remain unanswered. Are these adaptations mechanistically coupled? And, given that they

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service