Skip to Content
Merck
All Photos(1)

Key Documents

791113P

Avanti

18:1 DGS-NTA(Co)

1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (Cobalt salt), powder

Synonym(s):

1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (Cobalt salt), 1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] cobalt salt

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$548.00
10 MG
$877.00

$548.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$548.00
10 MG
$877.00

About This Item

Empirical Formula (Hill Notation):
C53H93CoN2O15
CAS Number:
Molecular Weight:
1057.24
UNSPSC Code:
12352211
NACRES:
NA.28

$548.00


Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

package of 1 × 10 mg (791113P-10MG)
package of 1 × 5 mg (791113P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COC(CCC(NCCCCC(C(O)=O)(N12(C3)CC(O1OO2C3=O)=O)[H])=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.O.[Co]

General description

1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (DGS-NTA) is a multivalent cationic lipid.[1]

Packaging

5 mL Clear Glass Sealed Ampule (791113P-5MG)
5 mL Clear Glass Sealed Ampule (791113P-10MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipid gene nanocarriers for the treatment of skin diseases: current state-of-the-art
Bellefroid C, et al.
European Journal of Pharmaceutics and Biopharmaceutics (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service