Skip to Content
Merck
All Photos(1)

Key Documents

780200P

Avanti

16:0 PE MCC

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidomethyl)cyclohexane-carboxamide] (sodium salt), powder

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidomethyl)cyclohexane-carboxamide] (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$1,260.00

$1,260.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 MG
$1,260.00

About This Item

Empirical Formula (Hill Notation):
C49H86N2O11PNa
CAS Number:
Molecular Weight:
933.18
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$1,260.00


Available to ship on22 April 2025Details


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (780200P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 780200P

shipped in

dry ice

storage temp.

−20°C

General description

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidomethyl)cyclohexane-carboxamide] (16:0 PE MCC) is a lipid consisting of phosphoethanolamine linked to two palmitic acid through its phosphate group and to a maleimide group by its amino group. It is a maleimide-functionalized lipid.[1]

Application

16:0 PE MCC (1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidomethyl)cyclohexane-carboxamide]) may been used in functionalized liposome preparation for drug delivery.[2] It may also be used in liposome preparation for testing drug delivery functionality[1] and as a component in microbubble dispersions.[3]

Biochem/physiol Actions

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidomethyl)cyclohexane-carboxamide](DPPE-MCC) via the maleimide group enables covalent attachment in functionalized liposomes and is useful in peptide transport studies.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (780200P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Astrid Hilde Myrset et al.
Ultrasound in medicine & biology, 37(1), 136-150 (2010-12-15)
Targeted ultrasound (US) contrast agents represent, because of their size (1 to 5 μm), a unique class of diagnostic imaging agents enabling true vascular imaging of conditions like inflammation and tumor angiogenesis. The objective of this study was to develop
K Gradauer et al.
Journal of controlled release : official journal of the Controlled Release Society, 172(3), 872-878 (2013-10-22)
The aim of the present study was the in vivo evaluation of thiomer-coated liposomes for an oral application of peptides. For this purpose, salmon calcitonin was chosen as a model drug and encapsulated within liposomes. Subsequently, the drug loaded liposomes
K Gradauer et al.
Journal of controlled release : official journal of the Controlled Release Society, 165(3), 207-215 (2012-12-12)
An ideal oral drug carrier should facilitate drug delivery to the gastrointestinal tract and its absorption into the systemic circulation. To meet these requirements, we developed a thiomer-coated liposomal delivery system composed of 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) and a maleimide-functionalized lipid, to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service