Skip to Content
Merck
All Photos(1)

Key Documents

700090P

Avanti

lanosterol-d6

Avanti Research - A Croda Brand

Synonym(s):

111016

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$769.00

$769.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$769.00

About This Item

Empirical Formula (Hill Notation):
C30H44D6O
CAS Number:
Molecular Weight:
432.75
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$769.00


Check Cart for Availability

Request a Bulk Order

description

26,26,26,27,27,27-hexadeuteriolanosta-8,24-dien-3β-ol

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700090P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

Application

Lanosterol-d6 has been used as an internal standard to quantify lanosterol by gas chromatography-mass spectrometry (GC-MS).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (700090P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Ester of ??Hydroxybutyrate Regulates Cholesterol Biosynthesis in Rats and a Cholesterol Biomarker in Humans
Kemper MF, et al.
Lipids, 50(12), 1185-1193 (2015)
Louise Ménégaut et al.
Cell reports, 31(7), 107665-107665 (2020-05-21)
Low-grade inflammation is constitutive of atherosclerosis, and anti-inflammatory therapy inhibiting interleukin-1β (IL-1β) reduces the rate of cardiovascular events. While cholesterol accumulation in atheroma plaque and macrophages is a major driver of the inflammatory process, the role of the LXR cholesterol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service