Skip to Content
Merck
All Photos(2)

Key Documents

700063P

Avanti

lanosterol

Avanti Research - A Croda Brand

Synonym(s):

lanosterin; lanosta-8,24-dienol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$315.00
5 MG
$1,080.00
10 MG
$1,940.00

$315.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$315.00
5 MG
$1,080.00
10 MG
$1,940.00

About This Item

Empirical Formula (Hill Notation):
C30H50O
CAS Number:
Molecular Weight:
426.72
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$315.00


Check Cart for Availability

Request a Bulk Order

description

8,24-lanostadien-3β-ol

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700063P-1mg)
pkg of 1 × 10 mg (700063P-10mg)
pkg of 1 × 5 mg (700063P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@H]1C([C@H]2[C@](CC1)(C3=C([C@]4([C@@]([C@H](CC4)[C@@H](CCC=C(C)C)C)(CC3)C)C)CC2)C)(C)C

InChI

1S/C30H50O/c1-20(2)10-9-11-21(3)22-14-18-30(8)24-12-13-25-27(4,5)26(31)16-17-28(25,6)23(24)15-19-29(22,30)7/h10,21-22,25-26,31H,9,11-19H2,1-8H3/t21-,22-,25+,26+,28-,29-,30+/m1/s1

InChI key

CAHGCLMLTWQZNJ-BQNIITSRSA-N

General description

Lanosterol is a lanostane-type tetracyclic triterpene,[1] which is the first sterol in lipid biosynthetic pathway.[2]

Biochem/physiol Actions

Lanosterol inhibits cataract formation, prevents lens opacity and reverses crystalline aggregation.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (700063P-10mg)
5 mL Amber Glass Screw Cap Vial (700063P-1mg)
5 mL Amber Glass Screw Cap Vial (700063P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lanosterol synthase pathway alleviates lens opacity in age-related cortical cataract
Shen X, et al.
Journal of Ophthalmology, 41(4), 744-747 (2018)
Tetracyclic triterpenes from Inonotus obliquus (PERS.) PIL.(CHAGA) growing in Russia
Zhukovich EN, et al.
Pharmaceutical Chemistry Journal, 2018, 495-496 (2010)
Lindsay M Webb et al.
The Journal of clinical investigation, 130(4), 1683-1698 (2020-02-25)
Protein arginine methyltransferase 5 (PRMT5) catalyzes symmetric dimethylation (SDM) of arginine, a posttranslational modification involved in oncogenesis and embryonic development. However, the role and mechanisms by which PRMT5 modulates Th cell polarization and autoimmune disease have not yet been elucidated.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service