Skip to Content
Merck
All Photos(1)

Key Documents

300213

Avanti

LNP Lipid Blend 2

Avanti Research - A Croda Brand

Synonym(s):

Lipid Blend 2 for LNP Formulation

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$363.00

$363.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$363.00

About This Item

UNSPSC Code:
12352211
NACRES:
NA.21

$363.00


Check Cart for Availability

Request a Bulk Order

form

solid

packaging

pkg of 1x1 ea (300213-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

Features and Benefits

Pre-mixed lipid blend composed of components suitable for producing nucleic acid-based lipid nanoparticles. Recommended for use with DNA/plasmid cargo.

Packaging

5 mL amber glass vial with aluminum seal (300213-1EA)

Preparation Note

Dissolve lipids in 1-mL ethyl alcohol. Use heat and sonication as necessary to achieve full dissolution. Dilute nucleic acid in aqueous buffer. Combine solvent and aqueous phase by solvent injection. Remove ethanol by dialysis.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • 890890P18:1 TAP (DOTAP), Avanti Research - A Croda BrandSDS

  • 700107PSitosterol (synthetic from plant sterol), Avanti Research - A Croda BrandSDS

  • 850365P18:0 PC (DSPC), Avanti Research - A Croda BrandSDS

  • 880151PDMG-PEG 2000, Avanti Research - A Croda BrandSDS

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service