300212AVT
Avanti
LNP Lipid Blend 1
Avanti Research™ - A Croda Brand
About This Item
Recommended Products
biological source
synthetic
Assay
>99% (TLC)
form
powder
packaging
pkg of 1X1 ea (300212-1EA)
manufacturer/tradename
Avanti Research™ - A Croda Brand
functional group
(Sphingolipid)
shipped in
dry ice
storage temp.
−20°C
Application
Features and Benefits
1) Dissolve cationic lipid in ethyl alcohol.
2) Once completely dissolved, use pipette to transfer cationic/ethanol solution into LNP Blend 1 vial.
3)Add amount of ethyl alcohol required to bring lipids to total desired volume. Use heat and sonication as necessary to achieve full dissolution. 4) Dilute nucleic acid in aqueous buffer (use appropriate pH for cationic lipid).
5) Combine solvent and aqueous phase by solvent injection. 6) Remove ethanol by dialysis.
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service