Skip to Content
Merck
All Photos(1)

Key Documents

Z755656

Microplate/tube head for Mortexer

pkg of 1 ea, for one microplate and microtubes (38 x 1.5 mL and 28 x 0.5 mL)

Synonym(s):

Optional horizontal head

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$90.70

$90.70


Check Cart for Availability


Select a Size

Change View
1 EA
$90.70

About This Item

UNSPSC Code:
41103811
NACRES:
NB.73

$90.70


Check Cart for Availability

sterility

non-sterile

packaging

pkg of 1 ea

manufacturer/tradename

Benchmark Scientific BV1000-COMBO

color

none

General description

Mortexer vortex head to hold microplate and tubes.

Legal Information

Mortexer is a trademark of Benchmark Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service