Skip to Content
Merck
All Photos(1)

Key Documents

Z742925

Solujet® detergent, low foaming, phosphate-free liquid

pkg of 19 L

Sign Into View Organizational & Contract Pricing

Select a Size

19 L
$521.00

$521.00


Check Cart for Availability


Select a Size

Change View
19 L
$521.00

About This Item

UNSPSC Code:
47131800
NACRES:
NB.34

$521.00


Check Cart for Availability

packaging

pkg of 19 L

manufacturer/tradename

Alconox, Inc.

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Solujet low-foaming phosphate-free liquid detergent

  • Concentrated to save you money
  • Phosphate-free, biodegradable and readily disposable
  • Low foaming high wetting detergent for maximum per formance
  • Free rinsing to give you reliable results and no interfering residues
  • Ideal for CIP cleaning and floor standing lab washing machines

Used to clean: Pharmaceutical process equipment, water testing labware, phosphate sensitive lab equipment cleaning, hospital instruments, optical components, pharmaceutical apparatus, cosmetics manufacturing equipment, tissue culture ware, plate glass, metal castings, forgings and stampings, industrial parts, animal cages, electronic components, tanks and reactors. USDA authorized category A1 for use in food contact surface cleaning. FDA certified.

Used to remove: Soil, radioisotopes, radioactivity, organics, grit, grime, slime, grease, tars, resins, fats, oils, blood, tissue, particulates, inorganic residues, chemical and solvents.

Surfaces cleaned: Recommended for glass, stainless steel, PTFE, ceramic, cement and alkali resistant plastics and metals such as stainless steel. Solujet is inhibited for Aluminum. Corrosion testing is advisable.
Clean-in place, laboratory warewashers, dishwashers, cart washers, spray washers, electrocleaning, ultrasonic cleaners, and manual cleaning if proper skin and eye protection is used

Directions: Follow machine manufacturers directions. Not intended for powder detergent receptacles. If available, use liquid detergent reservoir, connecting tubes or metering pump. In general, use 1/2 - 1% solution in hot water wash cycle (1 1/4 - 2 1/2 Tbsp. per gal., 3/4 - 1 1/4 oz. per gal. or 5-10 ml per liter--most machines need 2 1/2 oz. added at a typical 2 gal. wash cycle). For soak, circulate or ultrasonic use, make a 1% solution in cold, warm or hot water.
For difficult soils, raise water temperature and use more detergent. Wear protective gloves and eyewear when handling.
RINSE THOROUGHLY preferably with running water. For critical cleaning, do final or all rinsing in distilled, deionized or purified water. For food contact surfaces, rinse with potable water.

Legal Information

Solujet is a registered trademark of Alconox, Inc.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service