Key Documents
Z693545
2mag bioMIXcontrol MS4 control unit
AC/DC input 230 V AC, UK plug
Synonym(s):
laboratory stirrers, magnetic stir plates, magnetic stirrers, stirrers
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
Product Z693545 is not currently sold in your country. Contact Technical Service
material
stainless steel plate
feature
CE compliant
packaging
pack of 1 ea
manufacturer/tradename
2mag 98604
parameter
5 to 250 rpm speed (stirring)
5-250 rpm speed
plate size × W × L
× 200 mm × 110 mm
capacity
capacity (4 x bioMIXdrive 1)
General description
Efficient control unit, for 4 drives 2mag bioMIXdrive 1, developed for the hard daily lab use and warming-free continuous operation.
Control unit for the connection of 4 stirring drive bioMIXdrive 1, individual and independent speed setting for each stirring drive (Master-Slave-Technology), also ON/OFF function, synchronous speed mode for all drives, extended speed range from 5 to 250 rpm, 6 watts/stirring point without any own-generated heating effect, automatic power setting for the warming-free continuous operation e.g. in CO2-incubators, jerk-free stirring, even and smooth stirring also at slow speeds.
Digital display for speed setting, SoftStart for reliable and safe accelerating of stirring bar, QuickSet for quick setting of start- and maximum speed, storage of last working status when turning off, firm stainless steel housing, easy to clean.
Interfaces: 0-10 volts, 4-20 mA, RS 232, USB, Ethernet.
Choose from one of the most recent versions:
Certificates of Analysis (COA)