Skip to Content
Merck
All Photos(2)

Key Documents

Z677051

ART® low retention SoftFit L Reload System

maximum volume 1200 μL, Tip design ART 1200L, Hinged rack

Synonym(s):

aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip

Sign Into View Organizational & Contract Pricing

Select a Size

4 X 768 EA
$1,690.00

$1,690.00


Check Cart for Availability


Select a Size

Change View
4 X 768 EA
$1,690.00

About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
volume:
1200 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
Hinged rack
Tip design ART 1200L
packaging:
pack of 4800 ea (4 packs 768)
manufacturer/tradename:
Molecular BioProducts 2789-05-HR

$1,690.00


Check Cart for Availability

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier
Hinged rack
Tip design ART 1200L

packaging

pack of 4800 ea (4 packs 768)

manufacturer/tradename

Molecular BioProducts 2789-05-HR

technique(s)

PCR: suitable

maximum volume

1200 μL

volume

1200 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Designed to be compatible with LTS, the Molecular Bio Products Softfit L Reload System provides ART barrier pipette tips in color coded inserts by volume size. Choose from five pipette tip volume sizes with the option of low retention technology.

Each individual insert within the Softfit L Reload System is clearly identified with both product number and lot number allowing researchers to enjoy space-saving and environmental benefits of a reload tip system without loosing product traceability.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.
SoftFit-L is a trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service