Z647535
Pyrex® measuring cylinder with spout and detachable plastic foot, complete
volume 100 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
graduations 1.0 mL
volume measuring range
100 mL
H
270 mm
Looking for similar products? Visit Product Comparison Guide
General description
- Complies with ISO 4788
- Glass cylinder has blue enamel graduations
- Polyethylene foot absorbs impact and reduces breakage
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service