Key Documents
Z612014
Julabo® bath tubing
metal (triple insulated with connector M16x1), L 3 m
Select a Size
1 EA
$2,020.00
$2,020.00
Check Cart for Availability
Select a Size
Change View
1 EA
$2,020.00
About This Item
UNSPSC Code:
41105108
NACRES:
NB.77
$2,020.00
Check Cart for Availability
material
metal (triple insulated with connector M16x1)
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8930214
parameter
−100-350 °C temperature
L
3 m
General description
Flexible connection of the application to the temperature control instrument for optimal use in the respective temperature range.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)