Skip to Content
Merck
All Photos(1)

Documents

Z568597

Krytox® XHT-BDZ grease

weight 2 oz (tube)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
15121900
NACRES:
NB.35

manufacturer/tradename

Du Pont

weight

2 oz (tube)

General description

Designed for high temperature applications in the 288°C (550°F) range and higher. Chemically inert, non-flammable. Has low oil evaporation and is thickened with a non-melting thickener to allow use at temperatures above the melting point of common greases. White, creamy consistency. Useful temp range: 23 to 680°F with intermittent spikes greater than or equal to 752°F.

Legal Information

Krytox is a registered trademark of E. I. du Pont de Nemours and Company

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service