Key Documents
Z563129
J-KEM® digital temperature monitor
thermocouple type, T, temp. range -200-250 °C
About This Item
UNSPSC Code:
41112303
Product Z563129 is not currently sold in your country. Contact Technical Service
AC/DC input
120 V
feature
thermocouple type T
manufacturer/tradename
J-Kem
parameter
-200-250 °C temp. range
General description
Displays the temperature of an attached reaction or other piece of equipment on a bright LED display. No alarms. 0.1 °C resolution.
Legal Information
J-KEM is a registered trademark of J-KEM Electronics, Inc.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
  1. Does this instrument need to be calibrated or is it install-ready? Additionally, does it need to be connected to a computer in addition to the chiller?
    1 answer
    1. The controller comes pre-calibrated and can be used immediately. It does not require a computer connection, although the user has the option to connect it to a computer for data logging of time and temperature.
      Helpful?