BAB609010800
SP Bel-Art® Durac® Calibrated Thermocouple Thermometer
temp. range at -200-1370 °C, 2 PROBE (K/J)
Synonym(s):
Calibrated thermoelectric thermometer, H-B calibrated thermocouple thermometer for K or J type probes, Thermocouple temperature sensor
Select a Size
$511.00
Select a Size
About This Item
$511.00
Recommended Products
feature
2 PROBE (K/J)
packaging
pack of 1 ea
manufacturer/tradename
HB Instruments B60901-0800
parameter
temp. range at -200-1370 °C
Related Categories
General description
Features and Benefits
- Wide temperature range allows accurate monitoring at extreme temperatures
- Max/min/average temperature recording capability
- Data hold
- Includes two K-type beaded wire probes (-20/250°C capability); one is calibrated and serialized
- Software CD (where included) for real time data collection and logging via USB
- Bluetooth® app available for use with B60911-0200
- Probes for wider temperature ranges available (B60901-1200 & B60901-1300) or use alternatively purchased probes; see chart for probe types accepted by each unit
- Tested and calibrated in H-B’s exclusive triple accredited/registered ISO/IEC 17025:2017, A2LA accredited laboratory (Cert #2448.01), against equipment whose calibration is traceable to NIST
- Individually serialized, CE-marked Note: calibration applies to included probe and tested port only
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)