Skip to Content
Merck
All Photos(1)

Key Documents

Z308951

Quickfit® ground sleeve stirrer glands

shaft D 6 mm, joint: ST/NS 19/26

Synonym(s):

ST20/2

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$303.00

$303.00


Estimated to ship on16 April 2025



Select a Size

Change View
1 EA
$303.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.72

$303.00


Estimated to ship on16 April 2025


Joint

joint: ST/NS19/26

manufacturer/tradename

Quickfit ST 20/2

availability

not available in USA and Canada

shaft D

6 mm

fitting

13 thread

Looking for similar products? Visit Product Comparison Guide

General description

Shaft that passes through screw thread adapterr in top of gland. Will operate at up to 700rpm and down to 0.1 torr. Cup shaped lubricant reservoir surrounds sleeve such that lubricant also acts as coolant.

Legal Information

Quickfit is a registered trademark of SciLabware Limited

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service