Key Documents
Z304840
Quickfit® drying tube
joint: ST/NS 24/29
Synonym(s):
MF23/3
Select a Size
1 EA
$122.00
$122.00
Check Cart for Availability
Select a Size
Change View
1 EA
$122.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$122.00
Check Cart for Availability
material
colorless tube
glass tube
Joint
joint: ST/NS24/29
sterility
non-sterile
feature
graduations: no
neck thread PUSH FIT (24/29 cone size)
manufacturer/tradename
Quickfit MF 23/3
availability
not available in USA and Canada
binding type
non-treated surface
General description
Angled. Retain desiccant with plug of glass wool.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)