Documents
Z300853
Quickfit® Reduction Adapters
top female joint: ST/NS 10/19, bottom male joint: ST/NS 14/23
Synonym(s):
DA01
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
Joint
female joint: ST/NS top10/19
male joint: ST/NS bottom14/23
manufacturer/tradename
Quickfit DA 01
availability
not available in USA and Canada
General description
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited