Key Documents
Z300977
Quickfit® Reduction Adapters
top female joint: ST/NS 24/29, bottom male joint: ST/NS 34/35
Synonym(s):
DA35
Select a Size
1 EA
$82.60
$82.60
Check Cart for Availability
Select a Size
Change View
1 EA
$82.60
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$82.60
Check Cart for Availability
Joint
female joint: ST/NS top24/29
male joint: ST/NS bottom34/35
manufacturer/tradename
Quickfit DA 35
availability
not available in USA and Canada
General description
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)