Key Documents
Z278416
Sigma-Aldrich
Heat transfer block
Three bowl-shaped recesses
Select a Size
1 EA
$112.00
$112.00
Check Cart for Availability
Select a Size
Change View
1 EA
$112.00
About This Item
UNSPSC Code:
41102404
NACRES:
NB.43
$112.00
Check Cart for Availability
feature
Holds 3 x flasks
manufacturer/tradename
Ace Glass 9594-16
Ace Glass 959416
General description
Aluminum blocks have excellent heat transfer rates and will not interfere with magnetic stirring. Replaces glass sand baths in microscale experiments. Use on top of stirring/hot plates.
Will fit 3 and 5 mL thin-walled vials and 5, 10, and 25 mL round-bottom flasks. Three bowl shaped recesses, one 20 mm straight-through hole, one deep bowl-shaped hole, and one hole for a thermometer. 31/2 in. square × 3/4 in. thick.
Choose from one of the most recent versions:
Certificates of Analysis (COA)